Anti-TOX2 Rabbit Polyclonal Antibody
PRSI27-264
- Antigen name:Thymocyte selection-associated high mobility group box 2
- Antigen symbol:TOX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:C20orf100|dJ1108D11.2|dJ495O3.1|GCX-1|GCX1
- Immunogen:A synthetic peptide corresponding a region of human C20orf100.
- Size:50 µg
- Pk:50 µG
This gene, which is located on chromosome 20, is predicted to encode for a signal protein with unknown function.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat