Anti-PGLS Rabbit Polyclonal Antibody
Catalog # PRSI27-180
Supplier: ProSci Inc.
Specifications
- Antigen name:6-phosphogluconolactonase
- Antigen symbol:PGLS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:6PGL|6PGL_Human
- Immunogen:A synthetic peptide corresponding a region of human PGLS.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
PGLS belongs to the glucosamine/galactosamine-6-phosphate isomerase family, 6-phosphogluconolactonase subfamily. It is implicated in the hydrolysis of 6-phosphogluconolactone to 6-phosphogluconate.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PGLS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human