Anti-GNAO1 Rabbit Polyclonal Antibody
Catalog # PRSI27-019
Supplier: ProSci Inc.
Specifications
- Antigen name:Guanine Nucleotide Binding Protein (G Protein), Alpha Activating Activity Polypeptide O
- Antigen symbol:GNAO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:DKFZp686O0962|G-ALPHA-o|GNAO|EIEE17
- Immunogen:A synthetic peptide corresponding a region of human GNAO1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Activated Goalpha interacted directly with PLZF, and enhanced its function as a transcriptional and cell growth suppressor. Goalpha might play a role in mediating extracellular signal-regulated kinase activation by G protein-coupled receptors in the brain.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GNAO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat