Anti-RAI14 Rabbit Polyclonal Antibody
PRSI26-991
- Antigen name:Retinoic Acid Induced 14
- Antigen symbol:RAI14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:RAI13|NORPEG
- Immunogen:A synthetic peptide corresponding a region of human RAI14.
- Size:50 µg
- Pk:50 µG
RAI14 contains 7 ANK repeats. The function of RAI14 remains unknown.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RAI14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat