Anti-SCCPDH Rabbit Polyclonal Antibody
Catalog # PRSI26-824
Supplier: ProSci Inc.
Specifications
- Antigen name:Saccharopine Dehydrogenase
- Antigen symbol:SCCPDH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CGI49|Saccharopine dehydrogenase|SCCPDH|SCPDH|CGI 49|Probable saccharopine dehydrogenase
- Immunogen:A synthetic peptide corresponding a region of human SCCPDH.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SCCPDH belongs to the saccharopine dehydrogenase family. The exact function of SCCPDH remains unknown.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SCCPDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human