Anti-STK32A Rabbit Polyclonal Antibody
Catalog # PRSI26-697
Supplier: ProSci Inc.
Specifications
- Antigen name:Serine/Threonine Kinase 32A
- Antigen symbol:STK32A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:A930015B13Rik|YANK1
- Immunogen:A synthetic peptide corresponding a region of human STK32A.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
STK32A belongs to the protein kinase superfamily, Ser/Thr protein kinase family. It contains 1 protein kinase domain. The function of the STK32A protein remains unknown.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: STK32A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human