Anti-CANT1 Rabbit Polyclonal Antibody
PRSI26-668
- Antigen name:Calcium Activated Nucleotidase 1
- Antigen symbol:CANT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:DBQD|SCAN1|SHAPY|SCAN-1
- Immunogen:A synthetic peptide corresponding a region of human CANT1.
- Size:50 µg
- Pk:50 µG
CANT1 belongs to the apyrase family.It is a calcium-dependent nucleotidase with a preference for UDP. The order of activity with different substrates is UDP > GDP > UTP > GTP. The enzyme has very low activity towards ADP and even lower activity towards ATP. And it does not hydrolyze AMP and GMP.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CANT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat