Anti-GSDMB Rabbit Polyclonal Antibody
PRSI25-777
- Antigen name:Gasdermin B
- Antigen symbol:GSDMB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:PRO2521|PP4052|GSDML
- Immunogen:A synthetic peptide corresponding a region of human GSDML.
- Size:50 µg
- Pk:50 µG
GSDML belongs to the gasdermin family.GSDML may play a role as secretory or metabolic product involved in secretory pathway. It may also play a role in achieving and maintaining the final differentiation state of epithelial cells.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GSDMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human