Anti-ZBTB3 Rabbit Polyclonal Antibody
PRSI25-536
- Antigen name:Zinc Finger and BTB Domain Containing 3
- Antigen symbol:ZBTB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Zinc finger and BTB domain-containing protein 3|ZBTB3
- Immunogen:A synthetic peptide corresponding a region of human ZBTB3.
- Size:50 µg
- Pk:50 µG
ZBTB3 contains 2 C2H2-type zinc fingers and 1 BTB (POZ) domain. It may be involved in transcriptional regulation.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZBTB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human