Anti-ZXDA Rabbit Polyclonal Antibody
PRSI25-460
- Antigen name:Zinc Finger, X-linked, duplicated A
- Antigen symbol:ZXDA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ZXDA.
- Size:50 µg
- Pk:50 µG
ZXDA contains 10 C2H2-type zinc fingers. It belongs to the ZXD family. ZXDA cooperates with CIITA to promote transcription of MHC class I and MHC class II genes.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZXDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human