Anti-Csnk1g1 Rabbit Polyclonal Antibody
PRSI25-379
- Antigen name:Casein kinase 1, gamma 1
- Antigen symbol:CSNK1G1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Casein kinase I isoform gamma-1|CK-I gamma1|CKI gamma 1|CSNK1G1|CK1G1|CK I gamma1|CKI- gamma1|CKI gamma-1|KC1G1|CKI-gamma-1|CKI-gamma 1|CKI gamma1
- Immunogen:A synthetic peptide corresponding a region of mouse Csnk1g1.
- Size:50 µg
- Pk:50 µG
CSNK1g1 is involved in regulation of fast synaptic transmission mediated by glutamate, the major excitatory neurotransmitter in the brain.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Csnk1g1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: