Specifications
- Antigen name:Zinc Finger Protein 471
- Antigen symbol:ZNF471
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ZNF471.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ZNF471 belongs to the krueppel C2H2-type zinc-finger protein family. Itcontains 15 C2H2-type zinc fingers and 1 KRAB domain. ZNF471 may be involved in transcriptional regulation.
ZNF471 antibody can be used for detection of ZNF471 by ELISA at 1:62500. ZNF471 antibody can be used for detection of ZNF471 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZNF471
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human