Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Clozapine, Reference Standards

Supplier:   USP
Clozapine, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1142107.
  • 1142108.
  • Description:
  • Clozapine
  • Clozapine Resolution Mixture
  • CAS No.:
  • 5786-21-0
  • 5786-21-0
  • Formula:
  • C₁₈H₁₉ClN₄
  • C₁₈H₁₉ClN₄
  • UN No.:
  • 2811
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1142107.
    Clozapine
    Clozapine
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - £619.00
    £619.00
    1 * 100 mg
       £619.00

    Specifications:
    More
    Specifications
    Cat. No.
    1142107.
    Description
    Clozapine
    CAS No.
    5786-21-0
    Formula
    C₁₈H₁₉ClN₄
    UN No.
    2811
  • 1142108.
    Clozapine Resolution Mixture
    Clozapine Resolution Mixture
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - £1,210.00
    £1,210.00
    1 * 30 mg
       £1,210.00

    Specifications:
    More
    Specifications
    Cat. No.
    1142108.
    Description
    Clozapine Resolution Mixture
    CAS No.
    5786-21-0
    Formula
    C₁₈H₁₉ClN₄
    UN No.
    2811