Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Budesonide, Reference Standards

Budesonide, Reference Standards

Supplier:   USP
Budesonide, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1078201.
  • 1078245.
  • 1078267.
  • 1078289.
  • Description:
  • Budesonide
  • Budesonide Related Compound E
  • Budesonide Related Compound G
  • Budesonide Related Compound L
  • Synonym(s):
  • 16α,17-[butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4,14-triene-3,20-dione
  • 16α,17-[butylidenebis(oxy)]-11β, 21-dihydroxypregna-4-ene-3,20-dione
  • 16α,17-[butylidenebis(oxy)]-21-hydroxypregna-1,4-diene-3,11,20-trione
  • CAS No.:
  • 51333-22-3
  • 131918-64-4
  • 137174-25-5
  • 216453-74-6
  • Formula:
  • C₂₅H₃₄O₆
  • C₂₅H₃₂O₆
  • C₂₅H₃₆O₆
  • C₂₅H₃₂O₆

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • Budesonide
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - £388.00
    £388.00
    1 * 200 mg
       £388.00

    Specifications:
    More
    Specifications
    Cat. No.
    1078201.
    Description
    Budesonide
    Synonym(s)
    CAS No.
    51333-22-3
    Formula
    C₂₅H₃₄O₆
  • 1078245.
    Budesonide Related Compound E
    Budesonide Related Compound E
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - £1,100.00
    £1,100.00
    1 * 25 mg
       £1,100.00

    Specifications:
    More
    Specifications
    Cat. No.
    1078245.
    Description
    Budesonide Related Compound E
    Synonym(s)
    16α,17-[butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4,14-triene-3,20-dione
    CAS No.
    131918-64-4
    Formula
    C₂₅H₃₂O₆
  • 1078267.
    Budesonide Related Compound G
    Budesonide Related Compound G
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - £1,120.00
    £1,120.00
    1 * 25 mg
       £1,120.00

    Specifications:
    More
    Specifications
    Cat. No.
    1078267.
    Description
    Budesonide Related Compound G
    Synonym(s)
    16α,17-[butylidenebis(oxy)]-11β, 21-dihydroxypregna-4-ene-3,20-dione
    CAS No.
    137174-25-5
    Formula
    C₂₅H₃₆O₆
  • 1078289.
    Budesonide Related Compound L
    Budesonide Related Compound L
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - £1,150.00
    £1,150.00
    1 * 25 mg
       £1,150.00

    Specifications:
    More
    Specifications
    Cat. No.
    1078289.
    Description
    Budesonide Related Compound L
    Synonym(s)
    16α,17-[butylidenebis(oxy)]-21-hydroxypregna-1,4-diene-3,11,20-trione
    CAS No.
    216453-74-6
    Formula
    C₂₅H₃₂O₆
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.