Thioridazine, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1662504.
- 1663008.
- Description:
- Thioridazine
- Thioridazine Hydrochloride
- CAS No.:
- 50-52-2
- 130-61-0
- Formula:
- C₂₁H₂₆N₂S₂
- C₂₁H₂₇ClN₂S₂
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1662504.ThioridazineSpecifications:
More
Specifications
Cat. No.1662504.DescriptionThioridazineCAS No.50-52-2FormulaC₂₁H₂₆N₂S₂ - 1663008.Thioridazine HydrochlorideThioridazine HydrochlorideUSP reference standardVial200 mgLog in for availability1 * 200 mg£353.00Specifications:
More
Specifications
Cat. No.1663008.DescriptionThioridazine HydrochlorideCAS No.130-61-0FormulaC₂₁H₂₇ClN₂S₂
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



