Anti-TRAF3 Rabbit Polyclonal Antibody
BSBTA00424
- Antibody type:Primary
- Antigen symbol:TRAF3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for TRAF3 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: TRAF3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat