Anti-CD1c Rabbit Polyclonal Antibody
Catalog # BSBTA01188-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:CD1c
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for CD1c detection. Tested with WB, Direct ELISA in Human.
Type: Primary
Antigen: CD1c
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human