Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ADA Rabbit Polyclonal Antibody
Anti-ADA Rabbit Polyclonal Antibody
Catalog # BSBTA00866-2
Supplier:  Boster Bio
Anti-ADA Rabbit Polyclonal Antibody
Catalog # BSBTA00866-2
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ADA
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Rat,
    Mouse
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for ADA detection. Tested with WB, IHC-P, ELISA(Cap) in Mouse;Rat.

Type: Primary
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat