Order Entry
United Kingdom
ContactUsLinkComponent
Anti-RND1 Rabbit Polyclonal Antibody
Anti-RND1 Rabbit Polyclonal Antibody
Catalog # BSBTA06347
Supplier:  Boster Bio
Anti-RND1 Rabbit Polyclonal Antibody
Catalog # BSBTA06347
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Rho-related GTP-binding protein Rho6
  • Antigen symbol:
    RND1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Rho-related GTP-binding protein Rho6(RND1) detection. Tested with WB in Human;Mouse.

Type: Primary
Antigen: RND1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse