Anti-Il16 Rabbit Polyclonal Antibody
BSBTA01635
- Antibody type:Primary
- Antigen name:Pro-interleukin-16
- Antigen symbol:Il16
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Pro-interleukin-16(Il16) detection. Tested with WB, ELISA in Mouse.
Type: Primary
Antigen: Il16
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse