Anti-Complement C9 Rabbit Polyclonal Antibody
Catalog # BSBTA01010-2
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Complement C9
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Complement C9 detection. Tested with WB, Direct ELISA in Human;Mouse.
Type: Primary
Antigen: Complement C9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse