Order Entry
United Kingdom
ContactUsLinkComponent
Quetiapine Related Compounds, Reference Standard
Quetiapine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Quetiapine Related Compounds, Reference Standard
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    111974-74-4
  • Synonym(s):
    11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepine dihydrochloride
  • Formula:
    C₁₇H₁₇N₃S.₂HCl
  • Description:
    Quetiapine Related Compound B
  • UN No.:
    3077

Specifications

Related Information

Frequently Bought Together