Order Entry
United Kingdom
ContactUsLinkComponent
Domestic sludge - metals, NIST-2781 standard
Domestic sludge - metals, NIST-2781 standard
Catalog # PROONIST-2781
CAS Number:  
Domestic sludge - metals, NIST-2781 standard
Catalog # PROONIST-2781
CAS Number:  
Specifications
  • Pk:
    40 g
  • Description:
    NIST-2781, domestic sludge - metals
Specifications
About this item
This standard reference material is intended to use in the evaluation of methods used for the analysis of sludges and other materials of a similar matrix. These are dried and pulverised domestic sludge.
A unit consists of 40 g of the dried material.
Certifications: Traceable to NIST and SRM numbers.