Impurity and API standards, 4-nitro-2-phenoxyphenol
Catalog # PROOMM0378.07
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:50 mg
- Standards:Reference standards
- CAS No.:70995-08-3
- Description:4-Nitro-2-phenoxyphenol
- Formula:C₁₂H₉NO₄
- Description:4-Nitro-2-phenoxyphenol