Impurity and API standard, 1-acetyl-4-(4-hydroxyphenyl)piperazine
Catalog # PROOMM0145.05
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standards
- CAS No.:67914-60-7
- Description:1-Acetyl-4-(4-hydroxyphenyl)piperazine
- Formula:C₁₂H₁₆N₂O₂
- Description:1-Acetyl-4-(4-hydroxyphenyl)piperazine