Order Entry
United Kingdom
ContactUsLinkComponent
 
Impurity and API standard, aniline hydrochloride
  PROOMM0093.17
 :  
Impurity and API standard, aniline hydrochloride
  PROOMM0093.17
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    142-04-1
  • Description:
    Aniline hydrochloride
  • Formula:
    C₆H₇N·ClH
  • Description:
    Aniline Hydrochloride