Fluorescent cell counters, Cellometer® X
Fluorescent cell counter with brightfield and red fluorescent channel, X1
Catalog # NEXCMX1
Supplier: NEXCELOM BIOSCIENCE
Jump to:
Product Details & Documents
The Cellometer X are PC-based cell imagers for fluorescent image capture and analysis.
- Cell count and viability in less than 30 seconds
- Ability to perform cell-based assays
- Cell size range 0,5 to 15 µm and sample volume 20 µl
Both can perform cell counting, viability, intensity and population analysis, for yeast concentration in brewing fermentations.
Delivery information: Supplied with the Cellometer® software.
Documents
1001233 Datasheet Cellometer X1
Datasheet
Specifications
- Model:X1
- Description:Fluorescent cell counter with brightfield and red fluorescent channel
- Seq:001
- Cat. No.:NEXCMX1
- Sample volume:20 µl
- Cell size:0,5 − 15 µm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambers
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambers
Product Information
- Catalog #AvailabilityPrice
- Laptop computer for Cellometer®
Product Information
- Catalog #AvailabilityPrice
- Validation solution for Cellometer® fluorescent instruments (100 tests) in a vialValidation solution for Cellometer® fluorescent instruments (100 tests) in a vialNEXCCCBM-011-2MLLog in for availability1 * 2 mL - £619.00
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambers
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambersDisposable plastic counting chambersNEXCCHT4-PD100-002Log in for availability1 * 100 items - £523.00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



