Chitosan, M.W. 50,000 - 190,000 Da, 20 - 300 cP (1% w/w CH₃COOH 1%), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9012-76-4
:MFCD00149018
MDL Number:MFCD00161512
:M-Clarity™ quality level = MQ100
Specification Test Results
- Appearance (color):Faint beige to beige
- Appearance (form):Powder and/or chips
- Deacetylation:≥75,0%
- Viscosity (c=1%, 1% acetic acid):20 - 300 cps
- Molecular weight:50,000 - 190,000 Da
- Solubility:Dilute aqueous acid: soluble
Specification Test Results
Frequently Bought Together