Perchloric acid 65 - 71%, NORMATOM®, Ultrapure for trace metal analysis
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7601-90-3
:
Formula:HClO₄
MDL Number:MFCD00011325
:100.46 g/mol
UN:1873
ADR:5.1,I
Specification Test Results
- Assay (HClO4):65 – 71% w/w
- Ag (Silver):max. 100 ppt
- Al (Aluminium):max. 100 ppt
- As (Arsenic):max. 100 ppt
- Ba (Barium):max. 100 ppt
- Be (Beryllium):max. 100 ppt
- Bi (Bismuth):max. 10 ppt
- Ca (Calcium):max. 100 ppt
- Cd (Cadmium):max. 10 ppt
- Ce (Cerium):max. 10 ppt
- Co (Cobalt):max. 100 ppt
- Cs (Cesium):max. 10 ppt
- Cu (Copper):max. 100 ppt
- Dy (Dysprosium):max. 10 ppt
- Er (Erbium):max. 10 ppt
- Eu (Europium):max. 10 ppt
- Fe (Iron):max. 100 ppt
- Ga (Gallium):max. 10 ppt
- Gd (Gadolinium):max. 10 ppt
- Hf (Hafnium):Information only
- Ho (Holmium):max. 10 ppt
- In (Indium):max. 10 ppt
- K (Potassium):max. 100 ppt
- La (Lanthanum):max. 10 ppt
- Li (Lithium):max. 100 ppt
- Lu (Lutetium):max. 10 ppt
- Mg (Magnesium):max. 100 ppt
- Mn (Manganese):max. 100 ppt
- Mo (Molybdenum):max. 100 ppt
- Na (Sodium):max. 100 ppt
- Nb (Niobium):Information only
- Nd (Neodymium):max. 10 ppt
- Ni (Nickel):max. 100 ppt
- Pb (Lead):max. 10 ppt
- Pd (Palladium):max. 10 ppt
- Pr (Praseodymium):max. 10 ppt
- Pt (Platinum):max. 100 ppt
- Rb (Rubidium):max. 10 ppt
- Rh (Rhodium):max. 10 ppt
- Sb (Antimony):max. 100 ppt
- Sc (Scandium):max. 100 ppt
- Sm (Samarium):max. 10 ppt
- Sn (Tin):max. 100 ppt
- Sr (Strontium):max. 100 ppt
- Ta (Tantalum):Information only
- Tb (Terbium):max. 10 ppt
- Te (Tellurium):max. 10 ppt
- Th (Thorium):max. 10 ppt
- Ti (Titanium):max. 100 ppt
- Tl (Thallium):max. 100 ppt
- Tm (Thulium):max. 10 ppt
- U (Uranium):max. 10 ppt
- V (Vanadium):max. 100 ppt
- W (Tungsten):Information only
- Y (Yttrium):max. 10 ppt
- Yb (Ytterbium):max. 10 ppt
- Zn (Zinc):max. 100 ppt
- Zr (Zirconium):max. 100 ppt
Specification Test Results
Related Information
Frequently Bought Together