Crucibles, quartz, self sealing
Catalog # 458-4221
Supplier: CEM
Specifications
- Capacity:10 ml
- Seq:01
- Environmentally Preferable:
- Pk:1
Specifications
About this item
This quartz crucible is self sealing and enables oxygen free ashing.
Delivery: Includes crucible and lid.