Anti-Substance P Mouse Monoclonal Antibody [clone: NC1/34]
BIRBORB233977-500
: Biorbyt
- Antibody type:Primary
- Antigen symbol:Substance P
- Clonality:Monoclonal
- Clone:NC1/34
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Pig
- Storage temperature:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at –20 °C, avoid freeze/thaw cycles.
- Immunogen:Full length
- Pk:500 µl
Type: Primary
Antigen: Substance P
Clonality: Monoclonal
Clone: NC1/34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Pig