Order Entry
United Kingdom
ContactUsLinkComponent
 
FluorSave™ reagent, Calbiochem®
  345789-20
 :  
FluorSave™ reagent, Calbiochem®
  345789-20
 :  

 

  • Pk:
    20 mL
  • Description:
    FluorSave™ reagent
  • Form:
    Liquid
  • Storage temperature:
    15 - 30 °C
  • Formulation:
    Formulated without glycerol

 

 

FluorSave™ reagent is an aqueous mounting medium for use in conjunction with fluorescent tracers such as fluorescein, rhodamine and phycobiliprotein probes.

  • Preserves fluorescence-stained tissue sections
  • Formulated without glycerol
  • Ideal for use with phycobiliprotein tracers