Rubber cement, fixogum, MP Biomedicals
Catalog # ICNA11FIXO0125
Supplier: MP Biomedicals
Specifications
- Description:Fixogum rubber cement, liquid
- Pk:125 mL
Specifications
About this item
Fixogum Rubber Cement is made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridisation for FISH.