Order Entry
United Kingdom
ContactUsLinkComponent
Restriction enzymes, Restriction enzyme, Eco147I (StuI), Fermentas
Restriction enzymes, Restriction enzyme, Eco147I (StuI), Fermentas
Catalog # ER0421
CAS Number:  
undefined
Restriction enzymes, Restriction enzyme, Eco147I (StuI), Fermentas
Catalog # ER0421
CAS Number:  

Specifications

  • Concentration:
    10 U/µl
  • Pk:
    1 KIT
  • Application:
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    Blunt
  • Source:
  • Description:
    Eco147I (StuI)
  • Environmentally Preferable:
  • Enzyme name:
    Eco147I (StuI)
  • Units:
    1000 units

Specifications

About this item

Thermo Scientific Eco147I (StuI) restriction enzyme recognises AGG^CCT sites and cuts best at 37 °C in B buffer (Isoschizomers: AatI, PceI, SseBI, StuI).

Applications include: Molecular cloning, restriction site mapping, genotyping, Southern blotting, restriction fragment length polymorphism (RFLP), SNP.

Kit is supplied as 3 vials: Eco1471 1000 units; 1 ml of 10X Buffer B; 1 ml of 10X Buffer Tango.