Water, Cell Culture Grade, tested to USP specifications for injection, sterile for sterile Water for Injection , Corning®
: Corning
: Hydrogen oxide
:7732-18-5
:
:H₂O
:MFCD00011332
:100
:18.02 g/mol
:0
:13,10098
:Ambient
- Environmentally Preferable:
- Mycoplasma:Tested negative
- Plating efficiency:Pass
- Oxidizable substances Solution:remains faintly pink
- Total organic carbon (before packaging) USP <643>:< 500 ppb
- Water conductivity (packaged water) USP <645>:Meets USP Requirements
- Water conductivity (before packaging) USP <645>:Meets USP temperature/conductivity requirements
- Endotoxin USP <85>:≤ 0.025 EU/mL
- Sterility USP <71>:Pass
- Particulate matter USP <788>:≤ 25 particles/mL that are ≥ 10 μm in size
≤ 3 particles/mL that are ≥ 25 μm in size
Frequently Bought Together