Order Entry
United Kingdom
ContactUsLinkComponent
XRF sample cup, 1500 series, Chemplex®
XRF sample cup, 1500 series, Chemplex®
Catalog # CHPXCH-1540
undefined
XRF sample cup, 1500 series, Chemplex®
Catalog # CHPXCH-1540
Specifications
  • Description:
    XRF sample cup, double open ends, 39,4 mm outside diameter, 39,4 mm max. O.D., aperture 31,0 mm, volume: 16 ml, 23,1 mm height
  • Pk:
    100
Specifications
About this item
This sample cups are need for containing and analysing samples by XRF Spectroscopy. Different series of sample cups are available. Sample cup holder compatability varies based on the different series.