Anti-RNF167 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-9232R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 167
- Antigen symbol:RNF167
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26001
- Antigen synonyms:RING105|E3 ubiquitin-protein ligase RNF167|RN167_HUMAN|RNF167.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May act as an E3 ubiquitin-protein ligase, or as part of the E3 complex, which accepts ubiquitin from specific E2 ubiquitin-conjugating enzymes, such as UBE2E1, and then transfers it to substrates, such as SLC22A18. May play a role in growth regulation involved in G1/S transition.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RNF167
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat