Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 9 Member 3
- Antigen symbol:SLC9A3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:isoform 3|Solute carrier family 9 sodium/hydrogen exchanger|NHE3|Solute carrier family 9 member 3|SLC9A 3|Slc9a3|MGC126718|Sodium/hydrogen exchanger|member 3|NHE-3|MGC126720.|Sodium/hydrogen exchanger 3|SL9A3_HUMAN|NHE 3|Na+/H+ exchanger 3|Sodium|apical epithelial
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
NHE-3 are integral membrane proteins that are expressed in most mammalian tissues, where they regulate intracellular pH and cell volume. NHEs mediate the transport of hydrogen (H+) ions out of cells in exchange for extracellular sodium (Na+) ions. While NHE-1 is ubiquitously expressed, the NHE isoforms 2-8 have distinct tissue- and cell type-dependent expression and inhibitory characteristics. NHE-3 localizes to the apical membrane of renal proximal tubules where it is responsible for most of the sodium transport and fluid reabsorption. NHE-3 translocates to internal pools where it mediates natriuresis when blood pressure increases abruptly. NHE-3 is also expressed in the stomach and functions to protect the mucosa by secreting protons that diffuse into the mucous cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC9A3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat