Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 9, Subfamily B (NHA1, Cation Proton Antiporter 1), Member 2
- Antigen symbol:SLC9B2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:133308
- Antigen synonyms:NHE10|NHEDC2|NHA2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Na+/H+ exchangers (NHEs) catalyze the transport of Na+ in exchange for H+ across membranes in organisms and are required for numerous physiological processes. NHEDC2 (Na+/H+ exchanger-like domain-containing protein 2), also known as NHA2, is a 537 amino acid mitochondrial protein. NHEDC2 is involved in organelle volume homeostasis by catalyzing the exchange of protons for Na+ and Li+ across the inner mitochondrial membrane. Found in red blood cells, NHEDC2 is required for bone resorption activity and osteoclast differentiation. As a multi-pass membrane protein, NHEDC2 is expressed as two isoforms produced by alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC9B2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat