Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ITM2A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-ITM2A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-9705R-A647
Supplier:  Bioss
Anti-ITM2A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-9705R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integral membrane Protein 2A
  • Antigen symbol:
    ITM2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9452
  • Antigen synonyms:
    E25A|BRICD2A
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    73-123/263
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The type II integral membrane (ITM2) protein family consists of three members ITM2A (also designated E25), ITM2B and ITM2C. ITM2A expression is high in osteogenic and lymphoid tissues, while both ITM2B and ITM2C are expressed in brain. Mutations in the ITM2B gene can lead to familial British dementia (fbd), and autosomal dominant disease characterized by progressive dementia, spasticity, and cerebellar ataxia, or familial Danish dementia (fdd), an autosomal dominant disorder characterized by cataracts, deafness, progressive ataxia, and dementia. The ITM2A 263-amino acid protein contains an N-terminal cytosolic domain, an uncleaved signal anchor sequence, and a tyrosine-rich C-terminal domain. Human ITM2A shares 91% homology with mouse ITM2A.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ITM2A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat