Specifications
- Antibody type:Primary
- Antigen name:Membrane-associated ring Finger (C3HC4)8, E3 ubiquitin protein ligase
- Antigen symbol:MARCH8
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:220972
- Antigen synonyms:MIR|Cellular modulator of immune recognition|RNF178|MARCH-VIII|MARCH8|E3 ubiquitin-protein ligase MARCH8|c-MIR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. MARCH8 (membrane-associated ring finger (C3HC4) 8), also known as MIR or RNF178 (RING finger protein 178), is a 291 amino acid multi-pass membrane protein that localizes to vesicle membranes and contains one RING-CH-type zinc finger. Expressed in a variety of tissues, including immature dendritic cells, MARCH8 functions as an E3 ubiquitin ligase that is thought to regulate immune responses by promoting the ubiquitination and subsequent degradation of target proteins, such as B7-2 and CD71.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MARCH8
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat