Anti-CKAP2 Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-7745R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:Cytoskeleton Associated Protein 2
- Antigen symbol:CKAP2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26586
- Antigen synonyms:TMAP|LB1|se20-10
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Possesses microtubule stabilizing properties. Involved in regulating aneuploidy, cell cycling, and cell death in a p53/TP53-dependent manner.Tissue specificity: Abundant in testis, thymus, and in tumor derived cell lines, while barely detectable in liver, prostate, and kidney.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CKAP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: