Anti-NCAPD3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-7734R-HRP
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Non-SMC Condensin II Complex, Subunit D3
- Antigen symbol:NCAPD3
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:23310
- Antigen synonyms:CAPD3|CAP-D3|hcp-6|hCAP-D3|hHCP-6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 and SMC4. hCAP-D3 is a regulatory non-SMC subunit of the condensin II complex.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: NCAPD3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat