Order Entry
United Kingdom
ContactUsLinkComponent
Anti-IKK alpha + IKK beta Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-IKK alpha + IKK beta Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-7557R-A488
 :  Bioss
Anti-IKK alpha + IKK beta Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-7557R-A488
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen symbol:
    IKK alpha + IKK beta
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

IKK is a serine protein kinase, and the IKK complex contains alpha and beta subunits (IKK alpha and IKK beta). IKK alpha and IKK beta interact with each other and both are essential for NFkB activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IKK alpha + IKK beta
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat