Anti-ACP5 Rabbit Polyclonal Antibody (Cy7®)
BOSSBS-6434R-CY7
: Bioss
- Antibody type:Primary
- Antigen name:Acid Phosphatase 5, tartrate resistant
- Antigen symbol:ACP5
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:54
- Antigen synonyms:SPENCDI|TRAP|HPAP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:100-150/325
- Purification:Purified by Protein A
- Pk:100 µl
Involved in osteopontin/bone sialoprotein dephosphorylation. Its expression seems to increase in certain pathological states such as Gaucher and Hodgkin diseases, the hairy cell, the B-cell, and the T-cell leukemias.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ACP5
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat