Anti-TWIST2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-4173R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Twist Family BHLH Transcription Factor 2
- Antigen symbol:TWIST2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:117581
- Antigen synonyms:AMS|SETLSS|DERMO1|bHLHa39|FFDD3|BBRSAY
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Twist2 (Twist homolog 2) is a basic helix-loop-helix (bHLH) transcription factor which acts as a transcriptional repressor. It binds to the E-box consensus sequence 5'-CANNTG-3' and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Efficient DNA binding requires dimerization with another bHLH protein. Twist2 inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TWIST2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat