Anti-CRTAM Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-4957R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Cytotoxic And Regulatory T Cell Molecule
- Antigen symbol:CRTAM
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:56253
- Antigen synonyms:CD355|CRTAM|Class-I MHC-restricted T-cell-associated molecule|Cytotoxic and regulatory T-cell molecule
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:40-90/393
- Purification:Purified by Protein A
- Pk:100 µl
Interaction with CADM1 promotes natural killer (NK) cell cytotoxicity and interferon-gamma (IFN-gamma) secretion by CD8+ cells in vitro as well as NK cell-mediated rejection of tumors expressing CADM3 in vivo.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CRTAM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity: