Anti-HIST3H3 Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-3186R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:Histone cluster 3 H3
- Antigen symbol:HIST3H3
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8290
- Antigen synonyms:H3.4|H3FT|H3t|H3/g
- Modification:phospho-Ser11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:5-45/136
- Purification:Purified by Protein A
- Pk:100 µl
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HIST3H3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat