Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-KEAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-3648R-FITC
 :  Bioss
Anti-KEAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-3648R-FITC
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Kelch-like ECH-associated protein 1
  • Antigen symbol:
    KEAP1
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9817
  • Antigen synonyms:
    KLHL19|KIAA0132|Cytosolic inhibitor of Nrf2|KEAP1|Kelch-like ECH-associated protein 1|INRF2|INrf2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    75-125/624
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Acts as a substrate adapter protein for the E3 ubiquitin ligase complex formed by CUL3 and RBX1 and targets NFE2L2/NRF2 for ubiquitination and degradation by the proteasome, thus resulting in the suppression of its transcriptional activity and the repression of antioxidant response element-mediated detoxifying enzyme gene expression. Retains NFE2L2/NRF2 and may also retain BPTF in the cytosol. Targets PGAM5 for ubiquitination and degradation by the proteasome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KEAP1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat