Anti-HIST4H4 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-3826R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Histone cluster 4 H4
- Antigen symbol:HIST4H4
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:121504
- Antigen synonyms:H4FJ|HIST1H4H|H4/I|H4/M|H4FI|H4/K|H4FA|H4/G|HIST2H4B|H4F2|H4/O|HIST1H4L|H4/D|H4FC|H4/E|H4/B|H4FE|HIST1H4J|H4FH|H4FM|H4FO|H4FB|HIST1H4K|H4/N|H4/p|H4/J|HIST2H4|H4/C|HIST1H4F|H4FG|HIST1H4I|H4/A|H4FK|H4/H|H4FN|H4FD|HIST2H4A
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:60-100/103
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HIST4H4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat